Få mere at vide
N-Acetyl Dapsone-d8 (Major), TRC
Artikelnummer. 30310475
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30310475 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310475 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16843710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl Dapsone-d8
CAS 1215635-13-4
Molekylær formel C14H6D8N2O3S
Mængde 10 mg
Synonym N-[4-[(4-Aminophenyl)sulfonyl]phenyl]-acetamide-D8, Acetyldapsone-D8, MADDS-D8, Monoacetyldapsone-D8
InChI formel InChI=1S/C14H14N2O3S/c1-10(17)16-12-4-8-14(9-5-12)20(18,19)13-6-2-11(15)3-7-13/h2-9H,15H2,1H3,(H,16,17)/i2D,3D,4D,5D,6D,7D,8D,9D
SMIL O=S(C1=C([2H])C([2H])=C(NC(C)=O)C([2H])=C1[2H])(C2=C([2H])C([2H])=C(N)C([2H])=C2[2H])=O
IUPAC navn N-[4-(4-amino-2,3,5,6-tetradeuteriophenyl)sulfonyl-2,3,5,6-tetradeuteriophenyl]acetamide
Molekylvægt (g/mol) 298.39