Få mere at vide
N-(Acetyl-d3)-S-benzyl-L-cysteine, TRC
Artikelnummer. 30310874
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30310874 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310874 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1684271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(Acetyl-d3)-S-benzyl-L-cysteine
CAS 201404-15-1
Renhedsgrad noter HPLC
Molekylær formel C12 2H3 H12 N O3 S
Mængde 1 mg
Synonym L-Cysteine, N-(acetyl-d3)-S-(phenylmethyl)- (9CI), N-(Acetyl-d3)-S-benzyl-L-cysteine, N-(Acetyl-d3)-S-(phenylmethyl)-L-cysteine, (2R)-2-(Acetamido-2,2,2-d3)-3-(benzylthio)propanoic acid, N-Acetyl-S-benzyl-L-cysteine-D3 (acetyl-D3)
InChI formel InChI=1S/C12H15NO3S/c1-9(14)13-11(12(15)16)8-17-7-10-5-3-2-4-6-10/h2-6,11H,7-8H2,1H3,(H,13,14)(H,15,16)/t11-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])C(=O)N[C@@H](CSCc1ccccc1)C(=O)O
IUPAC navn (2R)-3-benzylsulfanyl-2-[(2,2,2-trideuterioacetyl)amino]propanoic acid
Molekylvægt (g/mol) 256.34
Formel vægt 256.0961
Procent renhed >95