Få mere at vide
N-Acetyl-d3-L-leucine-d10 N-Methyl-d3-amide, CDN
Artikelnummer. 30324658
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324658 0.01 g
30312969 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324658 Leverandør CDN Leverandørnr. CDND61170.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-L-leucine Methylamide D16
CAS 1276197-56-8
Molekylær formel C9 D16 H2 N2 O2
Mængde 0.01 g
Synonym N-Acetyl-d3-L-leucine-d10-N-methyl-d3-amide, N-Acetyl-d3-L-leucine-d10 N-Methyl-d3-amide, (2 S)-2-(Acetyl-d3-amino)-N,4-di(methyl-d3)pentanamide-2,3,3,4,5,5,5-d7
InChI formel InChI=1 S/C9H18N2O2/c1-6(2)5-8(9(13)10-4)11-7(3)12/h6,8 H,5H2,1-4H3,(H,10,13)(H,11,12)/t8-/m0/s1/i1D3,2D3,3D3,4D3,5D2,6 D,8 D
SMIL [2 H]C([2 H])([2 H])NC(=O)[C@@]([2 H])(NC(=O)C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn (2 S)-2,3,3,4,5,5,5-heptadeuterio-2-[(2,2,2-trideuterioacetyl)amino]-N,4-bis(trideuteriomethyl)pentanamide
Molekylvægt (g/mol) 202.35
Formel vægt 202.2373 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity