Få mere at vide
N-Acetyl-d3-glycine-d2 N-Methyl-d3-amide, CDN
Artikelnummer. 30322936
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322936 0.1 g 0.1g
30311411 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322936 Leverandør CDN Leverandørnr. CDND63420.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Acetamido-N-methylacetamide-D8
CAS 1219802-57-9
Molekylær formel C5 D8 H2 N2 O2
Mængde 0.1 g
Synonym N-Acetyl-d3-glycine-d2-N-methyl-d3-amide, 2-(Acetyl-d3-amino)-N-(methyl-d3)acetamide-2,2-d2, 2-(Acet-d3-amido)-N-(methyl-d3)acetamide-2,2-d2, N-(Acetyl-d3)-N'-(methyl-d3)glycinamide-d2, 2-Acetamido-N-methylacetamide-d8, 2-Acetamido-N-methylacetamide-D8
InChI formel InChI=1 S/C5H10N2O2/c1-4(8)7-3-5(9)6-2/h3H2,1-2H3,(H,6,9)(H,7,8)/i1D3,2D3,3D2
SMIL [2 H]C([2 H])([2 H])NC(=O)C([2 H])([2 H])NC(=O)C([2 H])([2 H])[2 H]
IUPAC navn 2,2,2-trideuterio-N-[1,1-dideuterio-2-oxo-2-(trideuteriomethylamino)ethyl]acetamide
Molekylvægt (g/mol) 138.194
Formel vægt 138.124 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity