Få mere at vide
N-Acetyl-d3-glycine-2,2-d2, CDN
Artikelnummer. 30314636
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314636 0.25 g
30319442 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314636 Leverandør CDN Leverandørnr. CDND69980.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetylglycine-D5 (N-acetyl,2,2-D5)
CAS 1219805-82-9
Molekylær formel C4 D5 H2 N O3
Mængde 0.25 g
Synonym N-Acetylglycine-d5, N-Acetyl-d3-glycine-2,2-d2, N-Acetylglycine-D5 (N-acetyl,2,2-D5), N-Acetylglycine-d5, 2-(Acet-d3-amido)acetic-2,2-d2 acid
InChI formel InChI=1 S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C(=O)NC([2 H])([2 H])C(=O)O
IUPAC navn 2,2-dideuterio-2-[(2,2,2-trideuterioacetyl)amino]acetic acid
Molekylvægt (g/mol) 122.134
Formel vægt 122.074 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity