Få mere at vide
N-Acetyl-d3 Adamantamine, TRC
Artikelnummer. 30319928
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30319928 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319928 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16805250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-d3 Adamantamine
CAS 1217704-63-6
Molekylær formel C12H16D3NO
Mængde 50 mg
Synonym 1-Acetamido-d3-adamantane, 1-Acetamido-d3-tricyclo[3.3.1.1(3,7)]decane, 1-Acetylamino-d3-adamantane, N-(1-Adamanthyl)acetamide-d3, N-1-Acetyl-d3-1-adamantanamine
InChI formel InChI=1S/C12H19NO/c1-8(14)13-12-5-9-2-10(6-12)4-11(3-9)7-12/h9-11H,2-7H2,1H3,(H,13,14)/i1D3
SMIL O=C(C([2H])([2H])[2H])NC12C[C@@]3([H])C[C@@]([H])(C2)C[C@](C1)([H])C3
IUPAC navn N-(1-adamantyl)-2,2,2-trideuterioacetamide
Molekylvægt (g/mol) 196.3