Få mere at vide
N-Acetyl-D-penicillamine, TRC
Artikelnummer. 17982115
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17982115 1 g
17954995 5 g
17960895 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17982115 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1873651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15537-71-0
Molekylær formel C7 H13 N O3 S
Molekylvægt (g/mol) 191.25
Synonym D-Valine, N-acetyl-3-mercapto-, N-Acetyl-D-Penicillamine, Valine, N-acetyl-3-mercapto-, D- (8CI), N-Acetyl-3-mercaptovaline, N-Acetyl-D-penicillamine, N-Acetylpenicillamine
IUPAC navn (2S)-2-acetamido-3-methyl-3-sulfanylbutanoic acid
SMIL CC(=O)N[C@@H](C(=O)O)C(C)(C)S
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-Penicillamine
Analyt- eller komponentnavne N-Acetyl-D-Penicillamine
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C7H13NO3S/c1-4(9)8-5(6(10)11)7(2,3)12/h5,12H,1-3H3,(H,8,9)(H,10,11)/t5-/m0/s1
Løsningstype Neat
Industry Type Forensics
Mængde 1 g
Formel vægt 191.0616