Få mere at vide
N-Acetyl-D,L-Aspartic Acid 2,3,3-D3, TRC
Artikelnummer. 30326348
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30326348 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326348 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16859710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-D,L-Aspartic Acid 2,3,3-D3
CAS 89829-69-6
Molekylær formel C6 D3 H6 N O5
Mængde 10 mg
Synonym Aspartic-2,3,3-d3 acid, N-acetyl- (7CI), 2-Acetamidobutanedioic-2,3,3-d3 acid, N-Acetyl-DL-aspartic-2,3,3-d3 acid, N-Acetyl-DL-aspartic-2,3,3-D3 acid
InChI formel InChI=1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)/i2D2,4D
SMIL [2H]C([2H])(C(=O)O)C([2H])(NC(=O)C)C(=O)O
IUPAC navn 2-acetamido-2,3,3-trideuteriobutanedioic acid
Molekylvægt (g/mol) 178.158
Formel vægt 178.067