missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Acetyl-D-glutamic Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | N-Acetyl-D-glutamic Acid |
| CAS | 19146-55-5 |
| Molekylær formel | C7 H11 N O5 |
| Mængde | 1 g |
| Synonym | D-Glutamic acid, N-acetyl-, Glutamic acid, N-acetyl-, D- (8CI), N-Acetyl-D-glutamic acid, D-N-Acetyl glutamic acid, (2R)-2-Acetamidopentanedioic acid |
| InChI formel | InChI=1S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m1/s1 |
| SMIL | CC(=O)N[C@H](CCC(=O)O)C(=O)O |
| IUPAC navn | (2R)-2-acetamidopentanedioic acid |
| Molekylvægt (g/mol) | 189.17 |
| Formel vægt | 189.0637 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?