Få mere at vide
N-Acetyl-D-glutamic-2,3,3,4,4-d5 Acid, TRC
Artikelnummer. 30313629
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30313629 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313629 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17824210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-glutamic-2,3,3,4,4-D5 Acid
CAS 14341-87-8
Molekylær formel C7 D5 H6 N O5
Mængde 10 mg
Synonym Glutamic-2,3,3,4,4-d5 acid, N-acetyl-, D- (8CI), N-Acetyl-D-glutamic-2,3,3,4,4-d5 acid, (2R)-2-Acetamidopentanedioic-2,3,3,4,4-d5 acid, N-Acetyl-D-glutamic-2,3,3,4,4-D5 acid
InChI formel InChI=1S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m1/s1/i2D2,3D2,5D
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])[C@@]([2H])(NC(=O)C)C(=O)O
IUPAC navn (2R)-2-acetamido-2,3,3,4,4-pentadeuteriopentanedioic acid
Molekylvægt (g/mol) 194.197
Formel vægt 194.095