Få mere at vide
N-Acetyl-D-glucosamine-d3, TRC
Artikelnummer. 30320912
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30320912 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320912 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17800210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-glucosamine-d3
CAS 77369-11-0
Molekylær formel C8H12D3NO6
Mængde 10 mg
Synonym 2-(Acetyl-d3-amino)-2-deoxy-D-glucose, 2-(Acetamido-d3)-2-deoxy-D-glucose, NSC 524344-d3
InChI formel InChI=1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1/i1D3
SMIL O[C@H]1[C@H](O)[C@@H](NC(C([2H])([2H])[2H])=O)C(O)O[C@@H]1CO
IUPAC navn 2,2,2-trideuterio-N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molekylvægt (g/mol) 224.23