Få mere at vide
N-Acetyl D-Alpha-Methyl DOPA Dimethyl Ether (+)-Alpha-Methylbenzylamine Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Acetyl D-alpha-Methyl DOPA Dimethyl Ether (+)-alpha-Methylbenzylamine Salt |
| CAS | 17772-88-2 |
| Molekylær formel | C22H30N2O5 |
| Mængde | 100 mg |
| Synonym | D-N-Acetyl-3-(3,4-dimethoxyphenyl)-2-methyl-alanine (+)-α-Methylbenzylamine, N-Acetyl D-α-Methyl DOPA Dimethyl Ether (+)-α-Methylbenzylamine Salt |
| InChI formel | InChI=1S/C14H19NO5.C8H11N/c1-9(16)15-14(2,13(17)18)8-10-5-6-11(19-3)12(7-10)20-4;1-7(9)8-5-3-2-4-6-8/h5-7H,8H2,1-4H3,(H,15,16)(H,17,18);2-7H,9H2,1H3/t14-;7-/m11/s1 |
| SMIL | COC1=CC(C[C@@H](C)([NH]C(C)=O)C(O)=O)=CC=C1OC.C[C@H2](C2=CC=CC=C2)N |
| IUPAC navn | (R)-2-acetamido-3-(3,4-dimethoxyphenyl)-2-methylpropanoic acid(R)-1-phenylethan-1-amine |
| Molekylvægt (g/mol) | 402.48 |
| Formel vægt | 402.49 |