Få mere at vide
N-Acetyl-D-Alanine-3,3,3-d3, CDN
Artikelnummer. 30327475
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327475 0.25 g -
30308784 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327475 Leverandør CDN Leverandørnr. CDND82600.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-alanine-3,3,3-D3
CAS 2714437-93-9
Molekylær formel C5 D3 H6 N O3
Mængde 0.25 g
Synonym N-Acetyl-D-alanine-3,3,3-D3, N-Acetyl-D-alanine-d3, (2 R)-2-Acetamido-3,3,3-trideuteriopropanoic acid, (R)-2-Acetamidopropionic-3,3,3-d3 acid
InChI formel InChI=1 S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m1/s1/i1D3
SMIL [2 H]C([2 H])([2 H])[C@@H](NC(=O)C)C(=O)O
IUPAC navn (2 R)-2-acetamido-3,3,3-trideuteriopropanoic acid
Molekylvægt (g/mol) 134.1483
Formel vægt 134.0771 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity