Få mere at vide
N-Acetyl-D-alanine-2,3,3,3-d4, TRC
Artikelnummer. 30313896
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30313896 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313896 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16796610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-D-alanine-2,3,3,3-d4
CAS 2708343-30-8
Molekylær formel C5 D4 H5 N O3
Mængde 10 mg
Synonym N-Acetyl-D-alanine-2,3,3,3-D4, N-Acetyl-D-alanine-d4, (2R)-2-Acetamido-2,3,3,3-tetradeuteriopropanoic acid, (R)-2-Acetamidopropionic-2,3,3,3-d4 acid
InChI formel InChI=1S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m1/s1/i1D3,3D
SMIL [2H]C([2H])([2H])[C@@]([2H])(NC(=O)C)C(=O)O
IUPAC navn (2R)-2-acetamido-2,3,3,3-tetradeuteriopropanoic acid
Molekylvægt (g/mol) 135.155
Formel vægt 135.083