Få mere at vide
N-Acetyl-5-hydroxytryptamine, TRC
Artikelnummer. 30011978
Skift visning
Mængde:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011978 1 g
30003965 100 mg
17940775 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011978 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1791601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1210-83-9
Molekylær formel C12 H14 N2 O2
Molekylvægt (g/mol) 218.2518
Synonym Acetamide, N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-, Acetamide, N-[2-(5-hydroxyindol-3-yl)ethyl]- (6CI,7CI,8CI), Serotonin, N-acetyl-, N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]acetamide, 5-Hydroxy-N-acetyltryptamine, 5-Hydroxymelatonin, N-Acetyl-2-(5-hydroxyindol-3-yl)ethylamine, N-Acetyl-5-hydroxytryptamine, N-Acetylserotonin, Normelatonin, O-Demethylmelatonin
IUPAC navn N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide
SMIL CC(=O)NCCc1c[nH]c2ccc(O)cc12
Tekniske data
Kemisk navn eller materiale N-Acetyl-serotonin
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C12H14N2O2/c1-8(15)13-5-4-9-7-14-12-3-2-10(16)6-11(9)12/h2-3,6-7,14,16H,4-5H2,1H3,(H,13,15)
Formel vægt 218.1055
Procent renhed >95%