Få mere at vide
N-Acetyl-4-aminobiphenyl-d5, TRC
Artikelnummer. 30316889
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30316889 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316889 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16817210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Acetyl-4-aminobiphenyl-d5
CAS 1217046-73-5
Molekylær formel C14H8D5NO
Mængde 10 mg
Synonym N-[1,1'-Biphenyl-d5]-4-ylacetamide, 4-(Acetylamino)biphenyl-d5, 4-Acetamidobiphenyl-d5, 4'-Phenylacetanilide-d5, NSC 227192-d5, NSC 3134-d5, NSC 65931-d5, p-Phenylacetanilide-d5
InChI formel InChI=1S/C14H13NO/c1-11(16)15-14-9-7-13(8-10-14)12-5-3-2-4-6-12/h2-10H,1H3,(H,15,16)/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2ccc(NC(=O)C)cc2
IUPAC navn N-[4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]acetamide
Molekylvægt (g/mol) 216.29
Formel vægt 216.1311