missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Acetyl-3,5-dinitro-L-tyrosine, 98%

Brand:  Alfa Aesar™ H63186.03

Additional Details : CAS Number : 20767-00-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469738

  • 835.10 DKK / 1g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

20767-00-4
313.222
CFKZKLOBRPCKTF-ZETCQYMHSA-N
1551646
C11H11N3O8
MFCD00024239
n-acetyl-3,5-dinitro-l-tyrosine, ac-3,5-dinitro-tyr-oh, s-2-acetamido-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, 2s-2-acetamido-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, ac-3,5-dinitro-tyr-oh., n-acetyl-3,5-dinitro-l-tyrosin, n-acetyl-4-hydroxy-3,5-dinitrophenylalanine #
CC(=O)NC(CC1=CC(=C(C(=C1)[N+](=O)[O-])O)[N+](=O)[O-])C(=O)O

Specifications

N-Acetyl-3,5-dinitro-L-tyrosine
C11H11N3O8
n-acetyl-3,5-dinitro-l-tyrosine, ac-3,5-dinitro-tyr-oh, s-2-acetamido-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, 2s-2-acetamido-3-4-hydroxy-3,5-dinitrophenyl propanoic acid, ac-3,5-dinitro-tyr-oh., n-acetyl-3,5-dinitro-l-tyrosin, n-acetyl-4-hydroxy-3,5-dinitrophenylalanine #
CC(=O)NC(CC1=CC(=C(C(=C1)[N+](=O)[O-])O)[N+](=O)[O-])C(=O)O
1551646
Solid
1g
20767-00-4
MFCD00024239
CFKZKLOBRPCKTF-ZETCQYMHSA-N
313.222
313.22
98%
Videos
SDS