Få mere at vide
N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide, Mikromol™
Artikelnummer. 17923476
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17923476 100 mg
17920896 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17923476 Leverandør Mikromol Leverandørnr. MM0061.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
CAS 110104-37-5
Analyt- eller komponentnavne N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide
Forsendelsestilstand Room Temperature
Molekylær formel C10 H13 N5 O4
Certificeringer/compliance ISO 17025
Synonym Aciclovir Imp. F (EP), Acyclovir USP Related Compound F, Acyclovir USP RC F, N-[9-[(2-Hydroxyethoxy)-methyl]-6-oxo-6,9-dihydro-1H-purin-2-yl]acetamide, Acyclovir Related Compound F, Aciclovir Impurity F
InChI formel InChI=1S/C10H13N5O4/c1-6(17)12-10-13-8-7(9(18)14-10)11-4-15(8)5-19-3-2-16/h4,16H,2-3,5H2,1H3,(H2,12,13,14,17,18)
SMIL CC(=O)NC1=Nc2c(ncn2COCCO)C(=O)N1
IUPAC navn N-[9-(2-hydroxyethoxymethyl)-6-oxo-1H-purin-2-yl]acetamide
Molekylvægt (g/mol) 267.24
Mængde 100 mg
Formel vægt 267.0968 g/mol