Få mere at vide
N-[(6-Chloropyridin-3-yl)methyl]methylamine, TRC
Artikelnummer. 30005456
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005456 5 g
30010014 500 mg
30006774 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005456 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3801455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120739-62-0
Molekylær formel C7 H9 Cl N2
Molekylvægt (g/mol) 156.61
Synonym 6-Chloro-N-methyl-3-pyridinemethanamine, 1-(6-Chloropyridin-3-yl)-N-methylmethanamine, 2-Chloro-5-(methylaminomethyl)pyridine, N-(6-Chloro-3-pyridylmethyl)-N-methylamine, N-[(6-Chloropyridin-3-yl)methyl]methylamine
IUPAC navn 1-(6-chloropyridin-3-yl)-N-methylmethanamine
SMIL CNCc1ccc(Cl)nc1
Tekniske data
Kemisk navn eller materiale Acetamiprid metabolite IM-1-4
Mængde 5 g
InChI formel InChI=1S/C7H9ClN2/c1-9-4-6-2-3-7(8)10-5-6/h2-3,5,9H,4H2,1H3
Formel vægt 156.0454