missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Chloro-2-pyridyl)thiourea, 97%, Thermo Scientific™

Catalog Number 15496147
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1310420-68-8
C6H6ClN3S
187.645
MFCD20727755
FXAVGDZZGZAWGV-UHFFFAOYSA-N
n-6-chloro-2-pyridyl thiourea, n-6-chloro-2-pyridinyl thiourea
98001126
(6-chloropyridin-2-yl)thiourea
C1=CC(=NC(=C1)Cl)NC(=S)N
This item is not returnable. View return policy
1310420-68-8
C6H6ClN3S
187.645
MFCD20727755
FXAVGDZZGZAWGV-UHFFFAOYSA-N
n-6-chloro-2-pyridyl thiourea, n-6-chloro-2-pyridinyl thiourea
98001126
(6-chloropyridin-2-yl)thiourea
C1=CC(=NC(=C1)Cl)NC(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 192°C to 196°C
Quantity 250 mg
Formula Weight 187.65
Percent Purity 97%
Chemical Name or Material N-(6-Chloro-2-pyridyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'