Få mere at vide
N-563, MedChemExpress
Beskrivelse
N-563 is an analogue of deoxyspergualin with an immunostimulating activity,it promotes resistance to Candida albicans infection in mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-563 |
| Fysisk form | Solid |
| CAS | 140686-92-6 |
| Farve | Off-White |
| Mængde | 5 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C15H29N5O5 |
| Opløselighedsinformation | DMSO : < 1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble or slightly soluble) |
| SMIL | O=C(NCCC(O)=O)CC(O)CNC(CCCCCCNC(N)=N)=O |
| Molekylvægt (g/mol) | 359.42 |
| Vis mere |