missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(5-Trifluoromethyl-2-pyridyl)thiourea, 97%

Brand:  Alfa Aesar™ H33731.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15446587

  • 306.00 DKK / 250mg

Estimated Shipment
15-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

329794-00-5
221.201
RZHVTDFWNLGTLJ-UHFFFAOYSA-N
18506835
C1=CC(=NC=C1C(F)(F)F)NC(=S)N
C7H6F3N3S
MFCD20921616
5-trifluoromethyl pyridin-2-yl thiourea, 5-trifluoromethylpyridin-2-yl-thiourea, 5-trifluoromethyl pyridin-2-ylthiourea, n-5-trifluoromethyl-2-pyridyl thiourea
[5-(trifluoromethyl)pyridin-2-yl]thiourea

Specifications

N-(5-Trifluoromethyl-2-pyridyl)thiourea
250mg
C7H6F3N3S
5-trifluoromethyl pyridin-2-yl thiourea, 5-trifluoromethylpyridin-2-yl-thiourea, 5-trifluoromethyl pyridin-2-ylthiourea, n-5-trifluoromethyl-2-pyridyl thiourea
C1=CC(=NC=C1C(F)(F)F)NC(=S)N
221.201
221.2
202°C to 206°C
329794-00-5
MFCD20921616
RZHVTDFWNLGTLJ-UHFFFAOYSA-N
[5-(trifluoromethyl)pyridin-2-yl]thiourea
18506835
97%
Videos
SDS Documents