Få mere at vide
N-[5-(Aminosulfonyl)-2-methylphenyl]-acetamide, TRC
Artikelnummer. 30318592
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318592 250 mg
30315746 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30318592 Leverandør Toronto Research Chemicals Leverandørnr. TRCA629355250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[5-(Aminosulfonyl)-2-methylphenyl]-acetamide
CAS 17485-44-8
Molekylær formel C9H12N2O3S
Mængde 250 mg
Synonym 5'-Sulfamoyl-O-acetotoluidide
InChI formel InChI=1S/C9H12N2O3S/c1-6-3-4-8(15(10,13)14)5-9(6)11-7(2)12/h3-5H,1-2H3,(H,11,12)(H2,10,13,14)
SMIL CC(NC1=CC([S](N)(=O)=O)=CC=C1C)=O
IUPAC navn N-(2-methyl-5-sulfamoylphenyl)acetamide
Molekylvægt (g/mol) 228.27
Formel vægt 228.06