missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-tert-Butylphenyl)-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65644.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469796

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD10401532

Specifications

N-(4-tert-Butylphenyl)-4-chloro-3-sulfamoylbenzamide
MFCD10401532
Solid
1g
366.86
95%
Videos
SDS