missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 230.35 |
Percent Purity | 95% |
Chemical Name or Material | N-(4-piperidin-1-ylbenzyl)cyclopropanamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'