Få mere at vide
N-4-Phenyl Alpha-Benzylidene-d5 Isobutyrylacetamide, TRC
Artikelnummer. 30421313
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30421313 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421313 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31955750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-4-Phenyl a-Benzylidene-d5 Isobutyrylacetamide
CAS 222412-73-9
Molekylær formel C19H14D5NO2
Mængde 50 mg
Synonym N-4-Phenyl A-Benzylidene-D5 Isobutyrylacetamide
InChI formel InChI=1S/C19H19NO2/c1-14(2)18(21)17(13-15-9-5-3-6-10-15)19(22)20-16-11-7-4-8-12-16/h3-14H,1-2H3,(H,20,22)/b17-13-/i3D,5D,6D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(\C=C(/C(=O)Nc2ccccc2)\C(=O)C(C)C)c([2H])c1[2H]
IUPAC navn (2Z)-4-methyl-3-oxo-2-[(2,3,4,5,6-pentadeuteriophenyl)methylidene]-N-phenylpentanamide
Molekylvægt (g/mol) 298.39
Formel vægt 298.173