Få mere at vide
N-[4-Nitro-3-(trifluoromethyl)phenyl]acetamide, Mikromol™
Artikelnummer. 30052568
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052568 25 mg
30052106 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052568 Leverandør Mikromol Leverandørnr. MM0161.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-Nitro-3-(trifluoromethyl)phenyl]acetamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[4-Nitro-3-(trifluoromethyl)phenyl]acetamide
CAS 393-12-4
Analyt- eller komponentnavne N-[4-Nitro-3-(trifluoromethyl)phenyl]acetamide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H7 F3 N2 O3
Certificeringer/compliance ISO 17025
Synonym Flutamide Imp. B (EP), Flutamide USP Related Compound B, Flutamide USP RC B, N-[4-Nitro-3-(trifluoromethyl)phenyl]acetamide, Flutamide Related Compound B, Flutamide Impurity B
InChI formel InChI=1S/C9H7F3N2O3/c1-5(15)13-6-2-3-8(14(16)17)7(4-6)9(10,11)12/h2-4H,1H3,(H,13,15)
SMIL CC(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC navn N-[4-nitro-3-(trifluoromethyl)phenyl]acetamide
Molekylvægt (g/mol) 248.16
Mængde 25 mg
Formel vægt 248.0409 g/mol