Få mere at vide
N-(4-Hydroxyphenyl)piperazine-d8, Dihydrobromide, TRC
Artikelnummer. 30405022
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30405022 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405022 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94946750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(4-Hydroxyphenyl)piperazine-d8, Dihydrobromide
CAS 1329502-88-6
Molekylær formel C10H8D8Br2N2O2
Mængde 50 mg
Synonym 4-(1-Piperazinyl)phenol-d8 Hydrochloride
InChI formel InChI=1S/C10H14N2O/c13-10-3-1-9(2-4-10)12-7-5-11-6-8-12/h1-4,11,13H,5-8H2/i5D2,6D2,7D2,8D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2ccc(O)cc2)C1([2H])[2H]
IUPAC navn 4-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)phenol
Molekylvægt (g/mol) 348.1
Formel vægt 186.1608