Få mere at vide
N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, CDN
Artikelnummer. 30314908
Skift visning
Mængde:
0.05 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314908 0.25 g
30314697 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314908 Leverandør CDN Leverandørnr. CDND63240.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetaminophen methyl-d3
CAS 60902-28-5
Molekylær formel C8 2H3 H6 N O2
Mængde 0.25 g
Synonym Acetaminophen D3 (Methyl D3), Acetamide-2,2,2-d3, N-(4-hydroxyphenyl)-, N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, Paracetamol D3
InChI formel InChI=1 S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11 H,1H3,(H,9,10)/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)Nc1ccc(O)cc1
IUPAC navn 2,2,2-trideuterio-N-(4-hydroxyphenyl)acetamide
Molekylvægt (g/mol) 154.18
Formel vægt 154.0822 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity