Få mere at vide
N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide-2,2,2-d3, TRC
Artikelnummer. 30322173
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322173 5 mg
30314210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322173 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1612335MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Paracetamol-D7
CAS 1219798-53-4
Molekylær formel C8 D7 H2 N O2
Mængde 5 mg
Synonym Acetamide-2,2,2-d3, N-(4-hydroxyphenyl-2,3,5,6-d4)-, Paracetamol-D4 (acetyl-D3,phenyl-D4), Paracetamol-D7, Paracetamol-d7, Acetaminophen-d7, 4-Hydroxy-2,3,5,6-tetradeutero(trideuteroacet)anilide, N-(4-Hydroxyphenyl-2,3,5,6-d4)acetamide-2,2,2-d3
InChI formel InChI=1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D
SMIL [2H]c1c([2H])c(NC(=O)C([2H])([2H])[2H])c([2H])c([2H])c1O
IUPAC navn 2,2,2-trideuterio-N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetamide
Molekylvægt (g/mol) 158.206
Formel vægt 158.107