Få mere at vide
N-[(4-Hydroxy-3-methoxyphenyl)methyl]octanamide (N-Vanillyloctanamide), Mikromol™
Artikelnummer. 17940786
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17940786 25 mg
17931826 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17940786 Leverandør Mikromol Leverandørnr. MM1033.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[(4-Hydroxy-3-methoxyphenyl)methyl]octanamide (N-Vanillyloctanamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide (N-Vanillyloctanamide)
CAS 58493-47-3
Analyt- eller komponentnavne N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide (N-Vanillyloctanamide)
Forsendelsestilstand Room Temperature
Molekylær formel C16 H25 N O3
Certificeringer/compliance ISO 17025
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-octanamide, N-Vanillyloctanamide
InChI formel InChI=1S/C16H25NO3/c1-3-4-5-6-7-8-16(19)17-12-13-9-10-14(18)15(11-13)20-2/h9-11,18H,3-8,12H2,1-2H3,(H,17,19)
SMIL CCCCCCCC(=O)NCc1ccc(O)c(OC)c1
IUPAC navn N-[(4-hydroxy-3-methoxyphenyl)methyl]octanamide
Molekylvægt (g/mol) 279.37
Mængde 25 mg
Formel vægt 279.1834 g/mol