missing translation for 'onlineSavingsMsg'
Få mere at vide

N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide (N-Vanillyldecanamide), Mikromol™

Artikelnummer. 17991196
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991196 100 mg
17982056 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17991196 Leverandør Mikromol Leverandørnr. MM1033.03

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide (N-Vanillyldecanamide)
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Vanillyldecanamide
CAS 31078-36-1
Analyt- eller komponentnavne N-Vanillyldecanamide
Forsendelsestilstand Room Temperature
Molekylær formel C18 H29 N O3
Certificeringer/compliance ISO 17025
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide, N-Vanillyldecanamide
InChI formel InChI=1S/C18H29NO3/c1-3-4-5-6-7-8-9-10-18(21)19-14-15-11-12-16(20)17(13-15)22-2/h11-13,20H,3-10,14H2,1-2H3,(H,19,21)
SMIL CCCCCCCCCC(=O)NCc1ccc(O)c(OC)c1
IUPAC navn N-[(4-hydroxy-3-methoxyphenyl)methyl]decanamide
Molekylvægt (g/mol) 307.43
Mængde 100 mg
Formel vægt 307.2147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.