Få mere at vide
N-(4'-Cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl] Urea, TRC
Artikelnummer. 30355391
Skift visning
Mængde:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355391 1 mg
30359073 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30355391 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9623451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1361414-26-7
Molekylær formel C26H18N6O
Molekylvægt (g/mol) 430.46
Synonym SR-9186
IUPAC navn 1-[4-(4-cyanophenyl)phenyl]-3-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl]urea
SMIL O=C(NC1=CC=C(C2=CC=C(C#N)C=C2)C=C1)NC(C=C3)=CC=C3C4=C5C(NC=N5)=NC=C4
Tekniske data
Kemisk navn eller materiale N-(4'-Cyano[1,1'-biphenyl]-4-yl)-N'-[4-(3H-imidazo[4,5-b]pyridin-7-yl)phenyl] Urea
Mængde 1 mg
InChI formel InChI=1S/C26H18N6O/c27-15-17-1-3-18(4-2-17)19-5-9-21(10-6-19)31-26(33)32-22-11-7-20(8-12-22)23-13-14-28-25-24(23)29-16-30-25/h1-14,16H,(H,28,29,30)(H2,31,32,33)
Formel vægt 430.1542