Få mere at vide
N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide, TRC
Artikelnummer. 30359597
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359597 100 mg
30365180 25 mg
30363032 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30359597 Leverandør Toronto Research Chemicals Leverandørnr. TRCC293005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 24234-92-2
Molekylær formel C17H16Cl2N2O3
Molekylvægt (g/mol) 367.23
Synonym 4'-Chloro-2'-(o-chlorobenzoyl)-2-[(2-hydroxyethyl)amino]acetamide, N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide
IUPAC navn N-[4-chloro-2-(2-chlorobenzoyl)phenyl]-2-(2-hydroxyethylamino)acetamide
SMIL OCCNCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2Cl
Tekniske data
Kemisk navn eller materiale N-[4-Chloro-2-(2-chlorobenzoyl)phenyl]-2-[(2-hydroxyethyl)amino]acetamide
Mængde 100 mg
InChI formel InChI=1S/C17H16Cl2N2O3/c18-11-5-6-15(21-16(23)10-20-7-8-22)13(9-11)17(24)12-3-1-2-4-14(12)19/h1-6,9,20,22H,7-8,10H2,(H,21,23)
Formel vægt 366.0538