Få mere at vide
N-[4-(-Carboxycyclohexylmethyl)]maleimide, TRC
Artikelnummer. 30363702
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363702 50 mg
30355158 250 mg
30364368 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363702 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17786050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64987-82-2
Molekylær formel C12 H15 N O4
Molekylvægt (g/mol) 237.25
Synonym Cyclohexanecarboxylic acid, 4-[(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)methyl]-, 4-[(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)methyl]cyclohexanecarboxylic acid, N-(4-Carboxycyclohexylmethyl)maleimide
IUPAC navn 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylic acid
SMIL OC(=O)C1CCC(CN2C(=O)C=CC2=O)CC1
Tekniske data
Kemisk navn eller materiale N-[4-(-Carboxycyclohexylmethyl)]maleimide
Mængde 50 mg
InChI formel InChI=1S/C12H15NO4/c14-10-5-6-11(15)13(10)7-8-1-3-9(4-2-8)12(16)17/h5-6,8-9H,1-4,7H2,(H,16,17)
Formel vægt 237.1001