Få mere at vide
N'-(4-Aminophenyl)-N,N-dimethylacetamidine-d6, TRC
Artikelnummer. 30327712
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30327712 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327712 Leverandør Toronto Research Chemicals Leverandørnr. TRCA62265210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N'-(4-Aminophenyl)-N,N-dimethylacetamidine-d6
CAS 1795133-90-2
Molekylær formel C10H9D6N3
Mængde 10 mg
Synonym N’-(4-Aminophenyl)-N,N-(dimethyl-d6)ethanimidamide, BAY-d 9216-d6, p-[1-(Dimethyl-d6)]aminoethylimino)aniline
InChI formel InChI=1S/C10H15N3/c1-8(13(2)3)12-10-6-4-9(11)5-7-10/h4-7H,11H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N(C(=Nc1ccc(N)cc1)C)C([2H])([2H])[2H]
IUPAC navn N'-(4-aminophenyl)-N,N-bis(trideuteriomethyl)ethanimidamide
Molekylvægt (g/mol) 183.28
Formel vægt 183.1643