Få mere at vide
N-(4-Amino-6,7-dimethoxyquinazol-2-yl)-N-(methyl-d3)-2-cyanoethylamine, TRC
Artikelnummer. 30313530
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30313530 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313530 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60496710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(4-Amino-6,7-dimethoxyquinazol-2-yl)-N-(methyl-d3)-2-cyanoethylamine
CAS 1185144-54-0
Molekylær formel C14H14D3N5O2
Mængde 10 mg
Synonym 3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)(methyl-d3)amino]propanenitrile
InChI formel InChI=1S/C14H17N5O2/c1-19(6-4-5-15)14-17-10-8-12(21-3)11(20-2)7-9(10)13(16)18-14/h7-8H,4,6H2,1-3H3,(H2,16,17,18)/i1D3
SMIL [2H]C([2H])([2H])N(CCC#N)c1nc(N)c2cc(OC)c(OC)cc2n1
IUPAC navn 3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-(trideuteriomethyl)amino]propanenitrile
Molekylvægt (g/mol) 290.34
Formel vægt 290.1571