Få mere at vide
N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide (Diacetylsulfadimidine; Diacetylsulfamethazine), Mikromol™
Artikelnummer. 30460756
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460756 4 x 25 mg
30052654 25 mg
30049084 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460756 Leverandør Mikromol Leverandørnr. MM1311.0800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide (Diacetylsulfadimidine; Diacetylsulfamethazine), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
CAS 59224-69-0
Analyt- eller komponentnavne N-[4-[Acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Forsendelsestilstand Room Temperature
Molekylær formel C16 H18 N4 O4 S
Certificeringer/compliance ISO 17025
Synonym N-[[4-(Acetylamino)phenyl]sulfonyl]-N-(4,6-dimethyl-2-pyrimidinyl)acetamide
InChI formel InChI=1S/C16H18N4O4S/c1-10-9-11(2)18-16(17-10)20(13(4)22)25(23,24)15-7-5-14(6-8-15)19-12(3)21/h5-9H,1-4H3,(H,19,21)
Løsningstype Neat
SMIL CC(=O)Nc1ccc(cc1)S(=O)(=O)N(C(=O)C)c2nc(C)cc(C)n2
IUPAC navn N-[4-[acetyl-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Molekylvægt (g/mol) 362.4
Mængde 4 x 25 mg
Formel vægt 362.1049 g/mol