missing translation for 'onlineSavingsMsg'
Learn More

N-[4,6-Dichloro-5-(formylamino)-2-pyrimidinyl]acetamide, TRC

Product Code. 30387177
Change view
Click to view available options
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30387177 250 mg
30387783 50 mg
30382705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387177 Supplier Toronto Research Chemicals Supplier No. TRCD434485250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136470-91-2
Molekylær formel C7 H6 Cl2 N4 O2
Molekylvægt (g/mol) 249.0541
Synonym Acetamide, N-[4,6-dichloro-5-(formylamino)-2-pyrimidinyl]-, N-(4,6-Dichloro-5-formamidopyrimidin-2-yl)acetamide, N-[4,6-Dichloro-5-(formylamino)-2-pyrimidinyl]acetamide
IUPAC navn N-(4,6-dichloro-5-formamidopyrimidin-2-yl)acetamide
SMIL CC(=O)Nc1nc(Cl)c(NC=O)c(Cl)n1

Specifications

Kemisk navn eller materiale N-(4,6-Dichloro-5-formamidopyrimidin-2-yl)acetamide
Mængde 250 mg
InChI formel InChI=1S/C7H6Cl2N4O2/c1-3(15)11-7-12-5(8)4(10-2-14)6(9)13-7/h2H,1H3,(H,10,14)(H,11,12,13,15)
Formel vægt 247.9868
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.