Få mere at vide
N-[4-[(5-Methylisoxazol-3-yl)sulphamoyl]phenyl]acetamide, Mikromol™
Artikelnummer. 30459998
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459998 4 x 25 mg
30049464 25 mg
17991486 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459998 Leverandør Mikromol Leverandørnr. MM0227.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-[(5-Methylisoxazol-3-yl)sulphamoyl]phenyl]acetamide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsulfamethoxazole
CAS 21312-10-7
Analyt- eller komponentnavne Acetylsulfamethoxazole
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 N3 O4 S
Certificeringer/compliance ISO 17025
Synonym Sulfamethoxazole Imp. A (EP), Sulfamethoxazole USP Related Compound A, Sulfamethoxazole USP RC A, N-[4-[(5-Methylisoxazol-3-yl)sulphamoyl]phenyl]acetamide, Sulfamethoxazole Related Compound A, Sulfamethoxazole Impurity A
InChI formel InChI=1S/C12H13N3O4S/c1-8-7-12(14-19-8)15-20(17,18)11-5-3-10(4-6-11)13-9(2)16/h3-7H,1-2H3,(H,13,16)(H,14,15)
Løsningstype Neat
SMIL CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2cc(C)on2
IUPAC navn N-[4-[(5-methyl-1,2-oxazol-3-yl)sulfamoyl]phenyl]acetamide
Molekylvægt (g/mol) 295.31
Mængde 4 x 25 mg
Formel vægt 295.0627 g/mol