Få mere at vide
N-[4-(4-Fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethenyl]-6-(1-methylethyl)pyrimidin-2-yl]-N-methylmethanesulfonamide (Rosuvastatin Lactone), Mikromol™
Artikelnummer. 17933626
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17933626 100 mg
17930576 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17933626 Leverandør Mikromol Leverandørnr. MM0936.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[4-(4-Fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethenyl]-6-(1-methylethyl)pyrimidin-2-yl]-N-methylmethanesulfonamide (Rosuvastatin Lactone)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rosuvastatin Lactone
CAS 503610-43-3
Analyt- eller komponentnavne Rosuvastatin Lactone
Forsendelsestilstand Room Temperature
Molekylær formel C22 H26 F N3 O5 S
Certificeringer/compliance ISO 9001
Synonym Rosuvastatin Calcium Imp. D (EP), Rosuvastatin Imp. D (EP), Methanesulfonamide, N-[4-(4-fluorophenyl)-6-(1-methylethyl)-5-[(1E)-2-[(2S,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethenyl]-2-pyrimidinyl]-N-methyl-, Rosuvastatin lactone, Rosuvastatin-5S-lactone, N-[4-(4-Fluorophenyl)-6-(1-methylethyl)-5-[(1E)-2-[(2S,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethenyl]-2-pyrimidinyl]-N-methylmethanesulfonamide, N-[4-(4-Fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethenyl]-6-(1-methylethyl)pyrimidin-2-yl]-N-methylmethanesulfonamide, Rosuvastatin Calcium Impurity D, Rosuvastatin Impurity D
InChI formel InChI=1S/C22H26FN3O5S/c1-13(2)20-18(10-9-17-11-16(27)12-19(28)31-17)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)32(4,29)30/h5-10,13,16-17,27H,11-12H2,1-4H3/b10-9+/t16-,17-/m1/s1
SMIL CC(C)c1nc(nc(c1\C=C\[C@@H]2C[C@@H](O)CC(=O)O2)c3ccc(F)cc3)N(C)S(=O)(=O)C
IUPAC navn N-[4-(4-fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxooxan-2-yl]ethenyl]-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
Molekylvægt (g/mol) 463.52
Mængde 100 mg
Formel vægt 463.1577 g/mol