Få mere at vide
N-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl] Rabeprazole Sulfide, TRC
Artikelnummer. 30466364
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466364 50 mg
30471595 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466364 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26517550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1114543-47-3
Molekylær formel C29 H36 N4 O4 S
Molekylvægt (g/mol) 536.69
Synonym 1-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]-2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole, Rabeprazole N-Alkyl Sulfide
IUPAC navn 1-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl]-2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfanyl]benzimidazole
SMIL COCCCOc1ccnc(CSc2nc3ccccc3n2Cc4nccc(OCCCOC)c4C)c1C
Tekniske data
Kemisk navn eller materiale N-[(4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl)methyl] Rabeprazole Sulfide
Mængde 50 mg
InChI formel InChI=1S/C29H36N4O4S/c1-21-24(30-13-11-27(21)36-17-7-15-34-3)19-33-26-10-6-5-9-23(26)32-29(33)38-20-25-22(2)28(12-14-31-25)37-18-8-16-35-4/h5-6,9-14H,7-8,15-20H2,1-4H3
Formel vægt 536.2457