Få mere at vide
N-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride, Mikromol™
Beskrivelse
N-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride |
| CAS | 1432053-72-9 |
| Analyt- eller komponentnavne | N-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H26 N2 O3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Acebutolol Hydrochloride Imp. E (EP), Acebutolol Imp. E (EP), N-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]butanamide Hydrochloride, Acebutolol Hydrochloride Imp. E (EP) as Hydrochloride, Acebutolol Hydrochloride Impurity E as Hydrochloride, Acebutolol Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C21H27N3.H3O4P/c1-17(2)24(18(3)4)15-13-21(16-22,19-10-6-5-7-11-19)20-12-8-9-14-23-20;1-5(2,3)4/h5-12,14,17-18H,13,15H2,1-4H3;(H3,1,2,3,4) |
| SMIL | Cl.CCCC(=O)Nc1ccc(OCC(O)CNC(C)C)cc1 |
| IUPAC navn | N-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]butanamide;hydrochloride |
| Vis mere |