Få mere at vide
N-(3-Methylbutyryl)glycine-2,2-d2, CDN
Artikelnummer. 30325933
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325933 0.01 g
30321450 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325933 Leverandør CDN Leverandørnr. CDND62300.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(3-Methylbutanoyl)glycine-2,2-d2
CAS 1219795-09-1
Molekylær formel C7 2H2 H11 N O3
Mængde 0.01 g
Synonym N-(3-Methyl-1-oxobutyl)glycine-2,2-d2, Isovalerylglycine-d2, N-Isovaleroylglycine-d2, N-Isovalerylglycine-d2, N-(3-Methylbutanoyl)glycine-2,2-D2
InChI formel InChI=1 S/C7H13NO3/c1-5(2)3-6(9)8-4-7(10)11/h5H,3-4H2,1-2H3,(H,8,9)(H,10,11)/i4D2
SMIL [2 H]C([2 H])(NC(=O)CC(C)C)C(=O)O
IUPAC navn 2,2-dideuterio-2-(3-methylbutanoylamino)acetic acid
Molekylvægt (g/mol) 161.2
Formel vægt 161.1021 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity