Få mere at vide
N-(3-Methylbenzyl)piperazine Dihydrochloride, LoGiCal
Artikelnummer. 30462483
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462483 10 mg
1 options
Artikelnummer. 30462483 Leverandør LoGiCal Leverandørnr. LGCFOR0586.07
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(3-Methylbenzyl)piperazine Dihydrochloride
CAS 5321-61-9
Analyt- eller komponentnavne N-(3-Methylbenzyl)piperazine Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C12 H18 N2 . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym N-(3-Methylbenzyl)piperazine Dihydrochloride
InChI formel InChI=1S/C12H18N2.2ClH/c1-11-3-2-4-12(9-11)10-14-7-5-13-6-8-14;;/h2-4,9,13H,5-8,10H2,1H3;2*1H
Løsningstype Neat
SMIL Cl.Cl.Cc1cccc(CN2CCNCC2)c1
IUPAC navn 1-[(3-methylphenyl)methyl]piperazine;dihydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 263.21
Mængde 10 mg
Formel vægt 262.1004 g/mol