Få mere at vide
N-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl Lansoprazole Sulfide, TRC
Artikelnummer. 17963275
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963275 25 mg
17933275 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963275 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33101525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1083100-27-9
Molekylær formel C25 H22 F6 N4 O2 S
Molekylvægt (g/mol) 556.52
Synonym 1-[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]-2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]thio]-1H-benzimidazole
IUPAC navn 1-[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methyl]-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridyl]methylsulfanyl]benzimidazole
SMIL Cc1c(CSc2nc3ccccc3n2Cc4nccc(OCC(F)(F)F)c4C)nccc1OCC(F)(F)F
Tekniske data
Kemisk navn eller materiale N-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl Lansoprazole Sulfide
Mængde 25 mg
InChI formel InChI=1S/C25H22F6N4O2S/c1-15-18(32-9-7-21(15)36-13-24(26,27)28)11-35-20-6-4-3-5-17(20)34-23(35)38-12-19-16(2)22(8-10-33-19)37-14-25(29,30)31/h3-10H,11-14H2,1-2H3
Formel vægt 556.1368