missing translation for 'onlineSavingsMsg'
Få mere at vide

N-(3-Mercapto-2-methylpropanoyl)glycine-d5, TRC

Artikelnummer. 30476218
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476218 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476218 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25400210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Kemisk navn eller materiale N-(3-Mercapto-2-methylpropanoyl)glycine-d5
CAS 1184993-97-2
Molekylær formel C6 2H5 H6 N O3 S
Mængde 10 mg
InChI formel InChI=1S/C6H11NO3S/c1-4(3-11)6(10)7-2-5(8)9/h4,11H,2-3H2,1H3,(H,7,10)(H,8,9)/i1D3,3D2
SMIL [2H]C([2H])([2H])C(C(=O)NCC(=O)O)C([2H])([2H])S
IUPAC navn 2-[[3,3,3-trideuterio-2-[dideuterio(sulfanyl)methyl]propanoyl]amino]acetic acid
Molekylvægt (g/mol) 182.25
Formel vægt 182.0773
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.