Få mere at vide
N-(3-Mercapto-2-methylpropanoyl)glycine-d5, TRC
Artikelnummer. 30467958
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30467958 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467958 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2540021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(3-Mercapto-2-methylpropanoyl)glycine-d5
CAS 1184993-97-2
Molekylær formel C6 2H5 H6 N O3 S
Mængde 1 mg
InChI formel InChI=1S/C6H11NO3S/c1-4(3-11)6(10)7-2-5(8)9/h4,11H,2-3H2,1H3,(H,7,10)(H,8,9)/i1D3,3D2
SMIL [2H]C([2H])([2H])C(C(=O)NCC(=O)O)C([2H])([2H])S
IUPAC navn 2-[[3,3,3-trideuterio-2-[dideuterio(sulfanyl)methyl]propanoyl]amino]acetic acid
Molekylvægt (g/mol) 182.25
Formel vægt 182.0773