Få mere at vide
N-(3-Isopropylamino-2-hydroxypropyl) Pindolol-d14, TRC
Artikelnummer. 30403665
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30403665 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403665 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8240422.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(3-Isopropylamino-2-hydroxypropyl) Pindolol-d14
CAS 1246815-15-5
Molekylær formel C11 H12 N2 O3
Mængde 2.5 mg
Synonym 4-[2-Hydroxy-3-[(1-methylethyl-d7)amino]propoxy]-alpha-[[(1-methylethyl-d7)amino]methyl]-1H-indole-1-ethanol
InChI formel InChI=1S/C11H12N2O3/c1-5-9(13)8(7(14)3-4-12)11(16)6(2)10(5)15/h5-6H,3,13H2,1-2H3
SMIL CC1C(=O)C(C)C(=O)C(=C1N)C(=O)CC#N
IUPAC navn 3-(2-amino-3,5-dimethyl-4,6-dioxocyclohexen-1-yl)-3-oxopropanenitrile
Molekylvægt (g/mol) 220.225
Formel vægt 220.085