Få mere at vide
N-(3-Indolylacetyl)glycine, TRC
Artikelnummer. 30011571
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011571 250 mg
30012101 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011571 Leverandør Toronto Research Chemicals Leverandørnr. TRCI628005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13113-08-1
Molekylær formel C12 H12 N2 O3
Molekylvægt (g/mol) 232.24
Synonym N-[2-(1H-Indol-3-yl)acetyl]glycine, N-(1H-Indol-3-ylacetyl)glycine, Indole-3-acetylglycine
IUPAC navn 2-[[2-(1H-indol-3-yl)acetyl]amino]acetic acid
SMIL OC(=O)CNC(=O)Cc1c[nH]c2ccccc12
Tekniske data
Kemisk navn eller materiale N-(3-Indolylacetyl)glycine
InChI formel InChI=1S/C12H12N2O3/c15-11(14-7-12(16)17)5-8-6-13-10-4-2-1-3-9(8)10/h1-4,6,13H,5,7H2,(H,14,15)(H,16,17)
Mængde 250 mg
Formel vægt 232.0848